missing translation for 'onlineSavingsMsg'
Learn More

Bacteriological Agar, Ultrapure

Product Code. 15835078 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Unit Size:
11339.8g
2268g
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15835078 1 lb. 454g
15845078 5 lb. 2268g
15855078 25 lb. 11339.8g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15835078 Supplier Thermo Scientific Supplier No. J10906.P2
Only null left
Add to basket
This item is not returnable. View return policy

Used as a gelling agent in preparation of bacteriological culture media.

Used extensively as a gelling agent in the preparation of bacteriological culture media. Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Superior transparency
  • Reliable reproducibility
  • Absence of inhibitors capable of hindering optimal development of microorganisms
  • Extraneous matter reduced to minimum
  • Format: White to light cream-colored powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
TRUSTED_SUSTAINABILITY

Specifications

Color White to light cream-colored
Form Powder
Grade Ultrapure
Product Type Agar
For Use With (Application) Used as a Gelling Agent in Preparation of Bacteriological Culture Media.
Packaging Type Plastic Bottle
Quantity 1 lb.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.