missing translation for 'onlineSavingsMsg'
Learn More

Thomil™ Bacteromil Antiseptic Hand Gel

Product Code. 16886033
Encompass_Preferred
Click to view available options
Quantity:
12 Bottles
2 Pk.
8 Bottles
Unit Size:
Pack of 12
Pack of 2
Pack of 8
This item is not returnable. View return policy
For external use only. Use biocides safely. Always read the label and product information before use. Do not use on people sensitive to any product components, or for extended periods of time. Do not mix with other products. Use with caution in children under two years. Highly flammable liquid and vapour. Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

Product Code. 16886033

Brand: Thomil™ HAHP062

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
For external use only. Use biocides safely. Always read the label and product information before use. Do not use on people sensitive to any product components, or for extended periods of time. Do not mix with other products. Use with caution in children under two years. Highly flammable liquid and vapour. Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

Thomil™ Bacteromil is a specially-formulated rinse-free friction antiseptic hand gel for healthy skin that is virucidal, bactericidal and levuricidal, and also efficacy-tested against coronavirus.

  • A biocidal hand gel for complete, safe disinfection of healthy skin
  • Synergistic mixture of alcohols and surfactants formulated to WHO recommendations with a neutral pH
  • Complies with UNE-EN 1500 (Antiseptic by Friction), UNE-EN 13624 (Levuricide) and UNE-EN 14476 (Virucide) standards
  • Quick drying with a smooth, non-sticky finish
  • Does not require rinsing or subsequent drying
  • Ideal for frequent use anywhere, especially where soap and water are not readily available

Specifications

Color Light Blue
Certifications/Compliance (Europe) UNE-EN 1500 (Antiseptic by friction); UNE-EN 13624 (Levuricide); UNE-EN 14476 (Virucide)
Composition Denatured Ethanol (70%), Triclosan (0.20%), Benzalkonium Chloride (0.16%)
pH Range Neutral
Quantity 8 Bottles
Solution Type Ethanolic
Volume (Metric) 500 mL
compliance-icons compliance-icons
Product Identifier
  • X8 500ml No Rinse Antiseptic Gel
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, quarternaryammonium compounds,ethandiol, cyclohexane
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.