missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ Casitone
Description
- An animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest
- This supplement provides a nutritional profile based on casein
- The specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate
- It has been successfully used for the production of Escherichia coli-based recombinant proteins
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifikationer
Specifikationer
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Production of Escherichia coli-based recombinant proteins |
| Product Line | Bacto |
| Quantity | 10 kg |
| Source | Animal Origin, Casein |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?