missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Neopeptone

Product Code. 16219751 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16219751 500 g 500g
16209751 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16219751 Supplier Gibco™ Supplier No. 211681
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation.

  • Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein
  • This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation
  • It has been used as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci
  • Also used with Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health application
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Applications of Bacto Neopeptone include:

  • Recommended for use in media for detection of fungi
  • Reported to provide nutrients for support of spirochetes and protozoa
  • Microbial fermentation
  • Production of vaccines for animal health applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Neopeptone
Format Bottle
For Use With (Application) Production of vaccines for animal health applications
Product Line Bacto
Quantity 500 g
Source Animal origin, bovine and porcine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.