missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ TC Lactalbumin Hydrolysate

Product Code. 16269871
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
This item is not returnable. View return policy

Product Code. 16269871

Brand: Gibco™ 259962

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An animal origin peptone from the enzymatically hydrolyzed protein portion of milk whey.

This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Gibco™ Bacto™ TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines
Product Line Bacto
Quantity 500 g
Source Animal Origin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.