missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Tryptose

Product Code. 16289751
Change view
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16289751 10 kg 10kg
16299751 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16289751 Supplier Gibco™ Supplier No. 211709
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties.

Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.

Features:

  • Reported as beneficial for cell culture applications
  • Animal origin (AO) nutritional supplement

Applications of Bacto Tryptose include:

  • Tryptose is very useful for cultivation of streptococci, pneumococci, meningococci, and other fastidious organisms.
  • Tryptose Peptone is often used as a biomass enhancer for recombinant Escherichia coli production.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Cell Culture
Product Line Bacto
Quantity 10 kg
Source Animal Origin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.