missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Baird-Parker Agar (ISO) Base (Dehydrated)

Product Code. 12721680
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12721680 500 g 500g
12731680 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12721680 Supplier Thermo Scientific™ Supplier No. CM1127B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate, enumerate, and identify Staphylococcus aureus in food samples with selective Baird-Parker Agar (ISO) Base.

Isolate, enumerate, and identify coagulase positive Staphylococcus aureus in food samples with selective Thermo Scientific™ Oxoid™ Baird Parker Agar (ISO) Base (Dehydrated) while adhering to ISO 6888-1:1999.

Baird Parker Agar contains carefully balanced selective inhibitory agents, glycine, lithium, and tellurite, to suppress the growth of most of the bacteria present in foods without inhibiting coagulase positive staphylococci. The presence of egg yolk emulsion and sodium pyruvate aid the recovery of damaged cells for presumptive identification of Staphylococcus aureus in food specimens.

Baird-Parker Agar base is widely recommended by national and international bodies for the isolation of Staphylococcus aureus1.
  • Easy interpretation of test results as Staphylococcus aureus forms gray/black shiny colonies with surrounding clear zones.
  • Adheres to ISO 6888-1:1999 standards.
Baird-Parker2 developed this agar medium from the tellurite-glycine formulation of Zebovitz et al 3. and improved its reliability in isolating coagulase-positive staphylococci from foods. Baird-Parker added egg yolk emulsion as a diagnostic agent and sodium pyruvate to protect damaged cells and aid their recovery4.

Egg yolk emulsion makes the medium cream/yellow and opaque. Coagulase-positive staphylococci reduce tellurite to form black/grey shiny colonies which, after 48 hours, produce clear zones around the colonies caused by the proteolysis of egg yolk. On further incubation, most strains of coagulase-positive staphylococci form opaque haloes around the colonies.

Some strains of Staphylococcus saprophyticus may also produce both clear zones and opaque haloes but can be distinguished by the longer incubation required5.

Colonies typical of Staphylococcus aureus , but without an egg yolk reaction, should also be tested for coagulase production6. Egg yolk reaction negative Staphylococcus aureus may occur in some food, especially cheese.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Baird-Parker Agar (ISO) Base (Dehydrated)
Form Powder
Product Type Agar
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.