missing translation for 'onlineSavingsMsg'
Learn More

J.T. Baker Baker-flex™, Cellulose PEI, J.T. Baker™

Product Code. 15598294 Shop All J.T.Baker Products
missing translation for 'orderingAttributeHoverText'
Quantity:
25 Sheets
missing translation for 'unitSize'
1 pack
This item is not returnable. View return policy

Product Code. 15598294

missing translation for 'mfr': J.T. Baker 447304

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Flexible TLC Sheet

Chemical Identifiers

CAS 9004-34-6

Specifications

Color White
Physical Form Crystal or Powder
Solubility Information Insoluble in water
Quantity 25 Sheets
Odor Odorless
Chemical Name or Material Cellulose PEI
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.