missing translation for 'onlineSavingsMsg'
Learn More

BAKER Silica Gel 40μm Flash Chromatography Packing, J.T. Baker™

Product Code. 10290051 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10290051 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10290051 Supplier Avantor J.T.Baker Supplier No. 702401
  • ONLINE EXCLUSIVE PRICE
    133.13 €
    114.35 € / 1kg
    Save 18.78 €
    14% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Poly Bottle
Quantity 1 kg
CAS 63231-67-4
Molecular Formula H2O3Si
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
SMILES O[Si](=O)O
IUPAC Name dihydroxy(oxo)silane
Molecular Weight (g/mol) 78.098
PubChem CID 14768
Chemical Name or Material BAKER Silica Gel 40μm Flash Chromatography Packing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.