missing translation for 'onlineSavingsMsg'
Learn More

J.T. Baker BAKERBOND spe™ Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns, J.T. Baker™

Product Code. 10299430 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
30/Box
50/Box
Unit Size:
Pack of 30
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10299430 30/Box Pack of 30
10381561 50/Box Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10299430 Supplier J.T. Baker Supplier No. 709007
  • ONLINE EXCLUSIVE PRICE
    418.00 €
    381.60 € / Pack of 30
    Save 36.40 €
    8% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Disposable Extraction Columns

Chemical Identifiers

CAS 126850-16-6

Specifications

Odor Odorless
Quantity 30/Box
Physical Form Solid
Solubility Information Practically Insoluble
Chemical Name or Material Aromatic Sulfonic Acid (C6H5SO3H) Disposable Extraction Columns
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.