missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bakuchiol

Product Code. 16491348
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491348 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491348 Supplier MedChemExpress Supplier No. HYN0235
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bakuchiol is a phytoestrogen isolated from the seeds of Psoralea corylifolia L; has anti-tumor effects.

Specifications

Content And Storage -20°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 256.38 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 10309-37-2
Product Type Natural Products
Purity 95.0%
Solubility DMSO : 62.5 mg/mL (243.78 mM∣Need ultrasonic)
Synonym (S)-(+)-Bakuchiol
Target p38 MAPK∣Autophagy
Quantity 5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.