missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Balicatib

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Balicatib is a potent and selective cathepsin K inhibitor (IC50 values are 1.4, 56 and 480 nM for human, rat and mouse cathepsin K, respectively). Exhibits >300-fold selectivity for cathepsin K over cathepsins L, B and S. Long term supratherapeutic dosing increases tissue levels of cathepsins L and B in rats.
Specifications
Specifications
| Chemical Name or Material | N-[1-(Cyanomethylcarbamoyl)cyclohexyl]-4-(4-propylpiperazin-1-yl)benzamide |
| CAS | 354813-19-7 |
| Quantity | 50 mg |
| Target | Synthetic Cathepsin Inhibitors |
| Molecular Formula | C23H33N5O2 |
| Formula Weight | 411.54 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction