Learn More
Tocris Bioscience™ Balicatib
GREENER_CHOICE
Potent and selective cathepsin K inhibitor
Brand:  Tocris Bioscience™ 5585/10
Encompass
Product Code. 15776946
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Balicatib is a potent and selective cathepsin K inhibitor (IC50 values are 1.4, 56 and 480 nM for human, rat and mouse cathepsin K, respectively). Exhibits >300-fold selectivity for cathepsin K over cathepsins L, B and S. Long term supratherapeutic dosing increases tissue levels of cathepsins L and B in rats.
Specifications
Specifications
N-[1-(Cyanomethylcarbamoyl)cyclohexyl]-4-(4-propylpiperazin-1-yl)benzamide
354813-19-7