missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bandmate™ Blot Tray Cover

Product Code. 17145822 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Tray Cover
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17145822 1 Tray Cover Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17145822 Supplier Invitrogen™ Supplier No. BW0050
  • ONLINE EXCLUSIVE PRICE
    232.00 €
    187.92 € / Pack of 1
    Save 44.08 €
    19% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Bandmate Blot Tray Cover is a replacement cover specially designed for use with the Bandmate Automated Western Blot Processor mini and midi incubation trays

This tray cover is required to protect blots and reagents during western blot processing.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description Bandmate Tray Cover
For Use With (Application) Western Blot
For Use With (Equipment) Invitrogen Bandmate Automated Western Blot Processor
Product Line Bandmate
Type Tray Cover
Quantity 1 Tray Cover
Shipping Condition Ambient
Dimensions (L x W x H) 21 cm x 12.7 cm x 0.953 cm
Weight 0.091 kg
Weight (English) 3.2 oz
Weight (Metric) 0.091 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.