missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bandmate™ Reagent Tubes
Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 tubes
500 tubes
Unit Size:
Pack of 100
Pack of 500
Descrizione
The tubes are required for dispensing primary and secondary antibodies into the Bandmate mini and midi incubation trays during the automated western blot processing procedure. Two reagent tubes are required for each blot.
Specifica
Specifica
| Color | Clear |
| Description | Bandmate Reagent Tubes, 100 Pack |
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | Invitrogen Bandmate Automated Western Blot Processor |
| Product Line | Bandmate |
| Type | Reagent Tube |
| Quantity | 100 tubes |
| Shipping Condition | Ambient |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto