missing translation for 'onlineSavingsMsg'
Learn More

BAPTA tetraethyl ester, 98+%

Product Code. 11465087
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465087 1 g 1g
11475087 5 g 5g
11485087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465087 Supplier Thermo Scientific Alfa Aesar Supplier No. B21971.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 73630-07-6 | C30H40N2O10 | 588.654 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73630-07-6
Molecular Formula C30H40N2O10
Molecular Weight (g/mol) 588.654
MDL Number MFCD00532663
InChI Key OLXCPQDFHUCXBA-UHFFFAOYSA-N
Synonym bapta tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetraethyl ester, tetraethyl 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetate, 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetic acid ethyl ester, ethyl 2-2-2-2-bis 2-ethoxy-2-oxidanylidene-ethyl amino phenoxy ethoxy phenyl-2-ethoxy-2-oxidanylidene-ethyl amino ethanoate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy-n-2-ethoxy-2-oxoethyl anilino acetate, ethyl 2-2-2-2-bis 2-ethoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-ethoxy-2-oxoethyl amino acetate
PubChem CID 339960
IUPAC Name ethyl 2-[2-[2-[2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy]ethoxy]-N-(2-ethoxy-2-oxoethyl)anilino]acetate
SMILES CCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC2=CC=CC=C2N(CC(=O)OCC)CC(=O)OCC

Specifications

Melting Point 98°C to 102°C
Quantity 1 g
Formula Weight 588.66
Percent Purity ≥98%
Chemical Name or Material BAPTA tetraethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.