missing translation for 'onlineSavingsMsg'
Learn More

BAPTA tetrasodium salt hydrate, 98%

Product Code. 11485906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485906 25 g 25g
11465906 1 g 1g
11475906 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485906 Supplier Thermo Scientific Alfa Aesar Supplier No. B22258.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 336624-09-0 | C22H20N2Na4O10 | 564.37 g/mol

BAPTA tetrasodium salt hydrateis a high quality and sensitive compound for calcium signaling studies(investigation of signal transduction and apoptotic cascades, and neuroscience research). BAPTA, Tetrasodium Salt is useful for spectrophotometric monitoring of extracellular Ca2+ levels, with very little sensitivity to pH. Has also been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BAPTA tetrasodium salt hydrateis a high quality and sensitive compound for calcium signaling studies(investigation of signal transduction and apoptotic cascades, and neuroscience research). BAPTA, Tetrasodium Salt is useful for spectrophotometric monitoring of extracellular Ca2+ levels, with very little sensitivity to pH. Has also been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336624-09-0
Molecular Formula C22H20N2Na4O10
Molecular Weight (g/mol) 564.37
MDL Number MFCD02683087
InChI Key ZWSMLJACYSGFKD-UHFFFAOYSA-J
Synonym bapta-na4, bapta tetrasodium, bapta tetrasodium salt, sodium 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, bapta, tetrasodium salt, bapta tetrasodium salt hydrate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrasodium salt, tetrasodium 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetate, 1,2-bis 2-aminophenoxy-ethane-n,n,n, n-tetraacetic acid tetrasodium salt, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrasodium salt hplc
PubChem CID 2734965
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O

Specifications

Quantity 25 g
Solubility Information Soluble in water.
IUPAC Name tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Formula Weight 564.37 (Anhydrous)
Percent Purity 98%
Chemical Name or Material BAPTA tetrasodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.