missing translation for 'onlineSavingsMsg'
Learn More

Baquiloprim-2,4-dione, TRC

Product Code. 30331333
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331333 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331333 Supplier Toronto Research Chemicals Supplier No. TRCB11800525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Baquiloprim-2,4-dione
CAS 83028-30-2
Molecular Formula C17H18N4O2
Quantity 25 mg
Synonym 5-[[8-(Dimethylamino)-7-methyl-5-quinolinyl]methyl]-2,4(1H,3H)-pyrimidinedione
InChI Formula InChI=1S/C17H18N4O2/c1-10-7-11(8-12-9-19-17(23)20-16(12)22)13-5-4-6-18-14(13)15(10)21(2)3/h4-7,9H,8H2,1-3H3,(H2,19,20,22,23)
SMILES CN(C)c1c(C)cc(CC2=CNC(=O)NC2=O)c3cccnc13
IUPAC Name 5-[[8-(dimethylamino)-7-methylquinolin-5-yl]methyl]-1H-pyrimidine-2,4-dione
Molecular Weight (g/mol) 310.35
Formula Weight 310.143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.