missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Barbed Elbow/Tee/Y-Connector Kit, HDPE

Product Code. 11714979 Shop All Masterflex Products
Change view
Click to view available options
Packaging Quantity:
Set of 1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Unit Size
11714979 Set of 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11714979 Supplier Masterflex™ Supplier No. 3151413
  • ONLINE EXCLUSIVE PRICE
    162.00 €
    151.18 € / Pack of 1
    Save 10.82 €
    6% Off
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Organize the fittings at your workbench with these convenient fittings kits

  • Designed for quick and easy assembly without tools.
  • Combine these simple and economical fittings with flexible tubing.
  • For use in low-pressure applications (generally less than 30psi).
  • Kit includes the following connectors:
    • Elbows: 1/16in., 3/32in., 1/8in., 5/32in., 3/16in., 1/4in., 5/16in.
    • Tees: 1/16in., 3/32in., 1/8in., 5/32in., 3/16in., 1/4in., 5/16in.
    • Y-connectors: 1/16in., 3/16in., 1/4in., 3/8in.

Specifications

Description Barbed elbow/tee/Y-connector kit.
Material HDPE
Packaging Quantity Set of 1
Includes Elbows: 1/16 in., 3/32 in., 1/8 in., 5/32 in., 3/16 in., 1/4 in., 5/16 in. Tees: 1/16 in., 3/32 in., 1/8 in., 5/32 in., 3/16 in., 1/4 in., 5/16 in. Y-connectors: 1/16 in., 3/16 in., 1/4 in., 3/8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.