missing translation for 'onlineSavingsMsg'
Learn More

Bardoxolone methyl, MedChemExpress

Product Code. 16445648
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16445648 10 mg 10mg
16436166 100 mg 100mg
16431817 50 mg 50mg
16421666 500 mg 500mg
16451076 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16445648 Supplier Medchem Express Supplier No. HY13324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bardoxolone methyl (NSC 713200; RTA 402; CDDO Methyl ester) is a synthetic triterpenoid compound with potential antineoplastic and anti-inflammatory activities, acting as an activator of the Nrf2 pathway and an inhibitor of the NF-κB pathway.

Bardoxolone methyl (NSC 713200

Specifications

Chemical Name or Material Bardoxolone methyl
CAS 218600-53-4
Color White
Purity Grade Notes Research
Molecular Formula C32H43NO4
Quantity 10 mg
Synonym RTA 402 NSC 713200 CDDO Methyl ester
Solubility Information DMSO : 25 mg/mL (49.44 mM; Need ultrasonic)
SMILES O=C1C(C#N)=C[C@@]2(C)[C@](CC[C@]([C@@]3(C)[C@@]4([H])[C@@]5([H])[C@@](CCC(C)(C)C5)(C(OC)=O)CC3)(C)C2=CC4=O)([H])C1(C)C
Molecular Weight (g/mol) 505.69
Formula Weight 505.69
Percent Purity 95.01%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.