missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bardoxolone methyl, MedChemExpress
Description
Bardoxolone methyl (NSC 713200
Specifications
Specifications
| Chemical Name or Material | Bardoxolone methyl |
| CAS | 218600-53-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H43NO4 |
| Quantity | 200 mg |
| Synonym | RTA 402 NSC 713200 CDDO Methyl ester |
| Solubility Information | DMSO : 25 mg/mL (49.44 mM; Need ultrasonic) |
| SMILES | O=C1C(C#N)=C[C@@]2(C)[C@](CC[C@]([C@@]3(C)[C@@]4([H])[C@@]5([H])[C@@](CCC(C)(C)C5)(C(OC)=O)CC3)(C)C2=CC4=O)([H])C1(C)C |
| Molecular Weight (g/mol) | 505.69 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?