missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Baricitinib

Product Code. 18706006
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18706006 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18706006 Supplier R&D Systems™ Supplier No. 7222/50
  • ONLINE EXCLUSIVE PRICE
    340.70 €
    305.55 € / 50mg
    Save 35.15 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent JAK1 and JAK2 inhibitor; also inhibits JAK3 and Tyk2

Baricitinib is a highly potent JAK1 and JAK2 inhibitor (IC50 values are 5.9 and 5.7 nM, respectively). Also inhibits Tyk2 (IC50 = 53 nM). Displays >100-fold selectivity for JAK1/2 over JAK3 (IC50 = 53 nM for JAK3). Binds AAK1, BIKE, GAK and MPSK1 in vitro (Kd values are 17.2, 39.8, 134.4 and 68.5 nM, respectively). Inhibits IL-6-induced STAT phosphorylation and subsequent pro-inflammatory chemokine (MPC-1) and cytokine (IL-17 and IL-22) production in PBMCs and T-cells. Displays anti-inflammatory and disease modifying effects in the rat adjuvant arthritis model. In silico modelling predicts inhibition of SARS-CoV-2 cell entry.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Ethylsulfonyl)-3-[4-(7 H-pyrrolo[2,3-d]pyrimidin-4-yl)-1 H-pyrazol-1-yl]-3-azetidineacetonitrile
CAS 1187594-09-7
Quantity 50 mg
Product Line Tocris
Target JAK Kinase Inhibitors
Molecular Formula C16H17N7O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.