missing translation for 'onlineSavingsMsg'
Learn More

Barium, 99+%, rods, approx. 2 cm diameter

Product Code. 10331484 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10331484 500 g 500g
10712973 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10331484 Supplier Thermo Scientific Acros Supplier No. 317865000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-39-3 | Ba | 137.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Gray
Melting Point 725.0°C
Boiling Point 1640.0°C
CAS Min % 99.0
CAS Max % 100.0
Quantity 500 g
Assay Percent Range 99+%
Form Rod
Linear Formula Ba
Merck Index 15, 960
Formula Weight 137.34
Percent Purity ≥99%
Chemical Name or Material Barium, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium in Paraffin oil
Signal Word
  • Danger
Hazard Category
  • Substance which, in contact with water, emits flammable gases Category 1
  • Aspiration toxicity Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: White mineral oil, Barium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.