missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, 99+%, for analysis

Product Code. p-3776849 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10266610

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+) diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Color White
pH 7 to 8.5 (at 5% 20°C)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.0 to 102% (Complexometry)
Linear Formula (CH3CO2)2Ba
Merck Index 15, 961
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 255.43
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.46
Density 2.4600g/mL
Chemical Name or Material Barium acetate, For analysis ACS
Show More Show Less
compliance-icons
Product Identifier
  • Barium acetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.