missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, ACS, 99.0-102.0%

Product Code. 11356598
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356598 5 kg 5kg
11366598 250 g 250g
11346598 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356598 Supplier Thermo Scientific Alfa Aesar Supplier No. 012198.A7
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol

Barium acetate is used as a chemical reagent, textile mordant and lubricant. It is used to prepare other acetates and barium carbonate. It acts as a mordant for printing textile fabrics. It is utilized for drying paints and varnishes. It serves as a catalyst in organic synthesis. It plays an important role in the separation of glycosaminoglycans. Further, it acts as a cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. In addition, it is used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+);diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline
Quantity 5 kg
Odor Odorless
Density 2.47 g/mL
UN Number UN1564
Merck Index 14,965
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 255.43
Percent Purity 99.0 to 102.0%
Grade ACS
Chemical Name or Material Barium acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.