missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, Puriss. p.a., ACS Reagent, ≥99%, Solstice

Product Code. 15613800 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
50 kg
Unit Size:
1kg
250g
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613800 50 kg 50kg
15603800 250 g 250g
15693790 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15613800 Supplier Honeywell-Fluka Supplier No. 3230550KG
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 100.0000%
Physical Form Crystalline
Quantity 50 kg
Packaging Fiberboard Box
Density 2.46 g/cm3
Linear Formula C6H5CH3
UN Number NONH for all modes of transport
Beilstein 3693411
IUPAC Name barium(2+) diacetate
Formula Weight 255.42
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Barium acetate, Puriss. p.a., ACS Reagent, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.