missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide, 99%, pure, anhydrous

Product Code. 15229826 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 15229826

Brand: Thermo Scientific Acros 315380500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For ultra trace analysis | CAS: 10553-31-8 | BaBr2 | 297.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10553-31-8
Molecular Formula BaBr2
Molecular Weight (g/mol) 297.16

Specifications

Melting Point 850.0°C
Quantity 50 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula BaBr2
Merck Index 15,964
Solubility Information Solubility in water: 104.1g/100mL (20°C). Other solubilities: soluble in methanol, almost insoluble in ethanol, ethyl acetate,, acetone, dioxane
Formula Weight 297.16
Percent Purity 99%
Grade Pure
Chemical Name or Material Barium bromide
compliance-icons
Product Identifier
  • Barium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Barium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.