missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide dihydrate, 99.3%

Product Code. 11326598
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326598 100 g 100g
11336598 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326598 Supplier Thermo Scientific Alfa Aesar Supplier No. 012197.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-28-8 | BaBr2H4O2 | 333.17 g/mol

Barium bromide dihydrate is used to purify radium and make zinc bromide. It also used  precursor to chemicals used in photograph.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-28-8
Molecular Formula BaBr2H4O2
Molecular Weight (g/mol) 333.17
MDL Number MFCD00149153
InChI Key HQQMRSGBIPFNSN-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dibromide
SMILES O.O.[Br-].[Br-].[Ba++]

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.30%
Solubility Information Very soluble in water,soluble in methanol
Formula Weight 333.17 (297.14 Anhydrous)
Density 3.58
Chemical Name or Material Barium bromide dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.