missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide, ultra dry, 99.998% (metals basis)

Product Code. p-7050126
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11303847

Brand: Thermo Scientific Alfa Aesar 035678.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10553-31-8

Barium bromide is a precursor to chemicals used in photography and to other bromides. Barium bromide was used to purify radium in a process of fractional crystallization devised by Marie Curie. Since radium precipitates preferentially in a solution of barium bromide, the ratio of radium to barium in the precipitate would be higher than the ratio in the solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10553-31-8
MDL Number MFCD00003444

Specifications

Melting Point 850°C
Color White
Physical Form Beads
Quantity 1 g
UN Number UN1564
Merck Index 14,968
Solubility Information It is soluble in water.
Formula Weight 297.14
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.781 g/mL
Assay (metals basis)
Chemical Name or Material Barium bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.