missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10060040 Shop All Fisher Chemical Products
Change view
Click to view available options
500g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10060040 500g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10060040 Supplier Fisher Chemical Supplier No. B/0350/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 1450°C
Melting Point 811°C
CAS 513-77-9
Color White
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula CBaO3
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES [Ba++].[O-]C([O-])=O
IUPAC Name barium(2+) carbonate
Molecular Weight (g/mol) 197.34
Formula Weight 197.35g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Barium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.