missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, ACS, 99.0-101.0%

Product Code. 11353288
Change view
Click to view available options
Quantity:
500 g
1 kg
2 x 1 kg
Unit Size:
1kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353288 2 x 1 kg 2kg
11363288 500 g 500g
11373288 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353288 Supplier Thermo Scientific Alfa Aesar Supplier No. 036236.D3
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 513-77-9 | CBaO3 | 197.34 g/mol

Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point (decomposition)
Melting Point 811°C
Color White
Physical Form Powder
Quantity 2 x 1 kg
Assay Percent Range 99.0 to 101.0%
Solubility Information Soluble in most acids,with the exception of sulfuric acid. It is insoluble in water and ethanol.
IUPAC Name barium(2+);carbonate
Formula Weight 197.35
Grade ACS
Odor Odorless
Appearance White powder
Density 4.43
Chemical Name or Material Barium carbonate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.