missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, Solstice

Product Code. 15681070
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15681070 50 g 50g
15671070 250 g 250g
15661070 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15681070 Supplier Honeywell-Fluka Supplier No. 1172950G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS reagent

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Quantity 50 g
Linear Formula BaCO3
UN Number NONH for all modes of transport
Beilstein 7045119
IUPAC Name barium(2+) carbonate
Formula Weight 197.34g/mol
Grade ACS/puriss. p.a.
Chemical Name or Material Barium Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.