missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride solution, 20%, Solstice

Product Code. 15694930
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15694930 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694930 Supplier Honeywell-Fluka Supplier No. 353235L
  • ONLINE EXCLUSIVE PRICE
    249.00 €
    225.10 € / 5 L
    Save 23.90 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

0.2

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

CAS 10361-37-2
Physical Form Liquid
Quantity 5 L
Packaging Plastic Bottle
Molecular Formula BaCl2
MDL Number MFCD00003445
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Ba++]
IUPAC Name barium(2+);dichloride
Molecular Weight (g/mol) 208.23
PubChem CID 25204
ChEBI CHEBI:63317
Formula Weight 208.23g/mol
Percent Purity 20%
Grade Volumetric
Concentration or Composition (by Analyte or Components) 20%
Chemical Name or Material Barium chloride solution
Show More Show Less
compliance-icons
Product Identifier
  • Barium chloride solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: barium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.