missing translation for 'onlineSavingsMsg'
Learn More

Barium ethoxide, 99.5% (metals basis), 10% w/v in ethanol

Product Code. 11338028
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 11338028

Brand: Thermo Scientific Alfa Aesar 036588.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2914-19-4 | C4H10BaO2 | 227.449 g/mol

It is used in nano-materials & thin films; catalysis & synthesis and optics & glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2914-19-4
Molecular Formula C4H10BaO2
Molecular Weight (g/mol) 227.449
MDL Number MFCD00050488
InChI Key GYIWFHXWLCXGQO-UHFFFAOYSA-N
Synonym barium diethanolate, diethoxybarium, barium 2+ ; ethanolate, ethanol, barium salt 9ci, barium 2+ bis ethoxide, barium 2+ ion bis ethoxide, barium ethoxide solution w/v in ethanol trace metals basis 25ml
PubChem CID 6451474
IUPAC Name barium(2+);ethanolate
SMILES CC[O-].CC[O-].[Ba+2]

Specifications

Color Yellow
Linear Formula Ba(OC2H5)2
Quantity 100 mL
UN Number UN3274
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 227.42
Percent Purity 99.5%
Concentration or Composition (by Analyte or Components) 10% w/v in ethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Barium ethoxide
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.