Learn More
Barium fluoride, ultra dry, 99.998% (metals basis)
Description
Barium Fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Barium Fluoride is used in spectroscopic components. Barium Fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Boiling Point | 2260°C |
| Melting Point | 1353°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 50 g |
| UN Number | UN1564 |
| Merck Index | 14,974 |
| Solubility Information | It is soluble in methanol,ethanol. Sparingly soluble in water. |
| Formula Weight | 175.36 |
| Percent Purity | 99.998% |
| Vis mere |
RUO – Research Use Only
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.