missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide Monohydrate, 98%, Honeywell Fluka™

Product Code. 15654120 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Poly Bottle
Unit Size:
25g
500g
Product Code. Quantity Packaging unitSize
15654120 500 g Poly Bottle 500g
15644120 25 g Poly Bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 15654120

Brand: Honeywell-Fluka 342386500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

>98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22326-55-2
Molecular Formula BaH4O3
Molecular Weight (g/mol) 189.356
MDL Number MFCD00149151
InChI Key GKQTUHKAQKWLIN-UHFFFAOYSA-L
Synonym barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
PubChem CID 16211219
IUPAC Name barium(2+);dihydroxide;hydrate
SMILES O.[OH-].[OH-].[Ba+2]

Specifications

Boiling Point 780°C
Color White
pH 13.1 (21g/L at 25°C)
Quantity 500 g
Linear Formula Ba(OH)2 · H2O
UN Number UN3262
Formula Weight 189.36g/mol
Percent Purity 98%
Packaging Poly Bottle
Appearance Powder or Crystals or Chunks
Density 3.743g/mL at 25°C
Chemical Name or Material Barium hydroxide monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.