missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide Octahydrate, TRC

Product Code. 30348259
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30348259 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30348259

Brand: Toronto Research Chemicals TRCB12900025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium Hydroxide Octahydrate
CAS 12230-71-6
Molecular Formula BaH18O10
Quantity 25 g
Synonym Barium Dihydroxide Octahydrate, Dihydroxybarium Octahydrate
InChI Formula InChI=1S/Ba.10H2O/h;10*1H2/q+2;;;;;;;;;;/p-2
SMILES O.O.O.O.O.O.O.O.O[Ba]O
IUPAC Name dihydroxybarium;octahydrate
Molecular Weight (g/mol) 315.46
Formula Weight 315.9952
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.