missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide solution, Volumetric, 0.05 M Ba(OH)2 (0.1N), Solstice

Product Code. 15664780 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15664780 1 L 1 L
15674780 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15664780 Supplier Honeywell-Fluka Supplier No. 350501L
 Restricted Item
Estimated Shipment: 22-06-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 0.05 M Ba(OH)2 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17194-00-2
Molecular Formula BaH2O2
Molecular Weight (g/mol) 171.341
InChI Key RQPZNWPYLFFXCP-UHFFFAOYSA-L
Synonym barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous
PubChem CID 6093286
ChEBI CHEBI:32592
IUPAC Name barium(2+);dihydroxide
SMILES [OH-].[OH-].[Ba+2]

Specifications

Physical Form Liquid
Quantity 1 L
Linear Formula Ba(OH)2
MDL Number MFCD00003443
UN Number NONH for all modes of transport
Formula Weight 171.34g/mol
Grade Volumetric
Chemical Name or Material Barium hydroxide solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.