missing translation for 'onlineSavingsMsg'
Learn More

Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis, Honeywell Fluka™

Product Code. 15653260 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
5 kg
6 x 500 g
25 kg
Unit Size:
25kg
500g
5kg
Pack of 6
Product Code. Quantity unitSize
15653260 25 kg 25kg
15663260 500 g 500g
15683260 6 x 500 g Pack of 6
15673260 5 kg 5kg
4 options
This item is not returnable. View return policy

Product Code. 15653260

Brand: Honeywell-Fluka 3112825KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99%,≥52% Ba basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
IUPAC Name barium(2+);dinitrate

Specifications

Melting Point 592°C (decomposition)(literature)
CAS Max % 100.0000%
Quantity 25 kg
Linear Formula (NH4)2SO4
UN Number UN1446
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
Formula Weight 261.34
Percent Purity >99%
Grade ACS Reagent
Packaging Fiberboard Box
Density 3.24 g/cm3
Chemical Name or Material Barium nitrate, Puriss. p.a., ACS Reagent, ≥99%, ≥52% Ba basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.