missing translation for 'onlineSavingsMsg'
Learn More

Barium perchlorate, anhydrous

Product Code. 11371768
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11371768

Brand: Thermo Scientific Alfa Aesar 011657.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13465-95-7 | BaCl2O8 | 336.219 g/mol

Barium perchlorate, anhydrous is used to make explosives. It is also used as desiccant, ribonuclease determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-95-7
Molecular Formula BaCl2O8
Molecular Weight (g/mol) 336.219
MDL Number MFCD00003440
InChI Key OOULUYZFLXDWDQ-UHFFFAOYSA-L
Synonym barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
PubChem CID 61623
IUPAC Name barium(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]

Specifications

Color Colorless
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 336.24
Odor Odorless
Sensitivity Hygroscopic
Density 3.2
Chemical Name or Material Barium perchlorate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.