missing translation for 'onlineSavingsMsg'
Learn More

Barium Peroxide, Extra Pure, SLR, Fisher Chemical

Product Code. 10672251 Shop All Fisher Chemical Products
Click to view available options
:
500g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10672251

Brand: Fisher Chemical B/1080/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 450°C
CAS 1304-29-6
Color Beige
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula BaO2
MDL Number 3454
InChI Key ZJRXSAYFZMGQFP-UHFFFAOYSA-N
SMILES [Ba++].[O-][O-]
IUPAC Name barium(2+) dioxidanediide
Molecular Weight (g/mol) 169.33
Formula Weight 169.33g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.