missing translation for 'onlineSavingsMsg'
Learn More

Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%

Product Code. 11356607
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356607 5 g 5g
11376607 50 g 50g
11366607 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356607 Supplier Thermo Scientific Alfa Aesar Supplier No. 000653.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-39-3 | Ba | 137.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Silver to Gray
Appearance Silver Gray pieces
Quantity 5 g
Odor Odorless
Assay Percent Range 99.2% (Metals basis)
Form Pieces
Chemical Name or Material Barium
compliance-icons
Product Identifier
  • Barium pieces
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use fire-extinguishing powder to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.