missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfide, 99.7% (metals basis)

Product Code. 11397577
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11397577

Brand: Thermo Scientific Alfa Aesar 012848.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21109-95-5 | BaS | 169.39 g/mol

Barium sulfide has different characters, so it is used in a variety of ways, such as in electronics, paint pigments, dehairing hides, flame retardant, luminous paints, and generating pure hydrogen sulfide. Barium sulfide can deviate X-rays and helps to give clearly image of the soft tissue. Barium sulfide is a precursor to other barium compounds and also used as a short wavelength emitters for electronic displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21109-95-5
Molecular Formula BaS
Molecular Weight (g/mol) 169.39
MDL Number MFCD00014192
InChI Key CJDPJFRMHVXWPT-UHFFFAOYSA-N
Synonym barium sulfide, thioxobarium
PubChem CID 88791
IUPAC Name barium(2+) sulfanediide
SMILES [S--].[Ba++]

Specifications

Melting Point 1200°C
Color Gray to Yellow
Physical Form Powder
Quantity 10 g
UN Number UN3134
Merck Index 14,995
Solubility Information Soluble in water but insoluble in alcohol.
Formula Weight 169.4
Percent Purity 99.7%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Density 4.25 g/mL
Assay (metals basis)
Chemical Name or Material Barium sulfide
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • Barium sulfide
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Faresætning
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
  • EUH031-Contact with acids liberates toxic gas.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplerende oplysninger
  • MIXTURE LIST-Contains : Barium sulfide

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.