missing translation for 'onlineSavingsMsg'
Learn More

Barium titanate(IV), 99%

Product Code. 10226400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10226400 2.5 kg Plastic bottle 2.5kg
10694892 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10226400 Supplier Thermo Scientific Acros Supplier No. 196860025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifica

Melting Point 1625.0°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 99%
Linear Formula BaTiO3
Merck Index 15, 996
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alkalies, slightly soluble in diluted acids, soluble in concentrated h2so4 and hf
Formula Weight 233.24
Percent Purity 99%
Loss on Ignition 0.5% max.
Packaging Plastic bottle
Specific Gravity 6.08
Density 6.0800g/mL
Chemical Name or Material Barium titanate(IV), 99%
Vedi altri risultati Mostra meno risultati
compliance-icons
Produktidentifikator
  • Barium titanate(IV)
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
Gefahrenhinweis
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.