Learn More
Barium titanium oxide, 99.7% (metals basis)
Description
Barium titanium oxide is a dielectric ceramic widely used in multi-layer ceramic capacitors (MLCCs), embedded decoupling capacitors (EDC) and ceramic filters. It is a piezoelectric and ferroelectric component used in microphones and transducers. It is also useful for thermistors and self-regulating electric heating systems. It is doped with small amounts of metals like scandium, yttrium, neodymium, samarium to become a semiconductor. It is used in some types of uncooled sensors for thermal cameras and in electric vehicles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1620°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Odor | Odorless |
| Density | 5.85 g/mL |
| Merck Index | 14,1000 |
| Solubility Information | Slightly soluble in dilute mineral acids. Soluble in concentrated hydrofluoric acid. Insoluble in water. |
| Formula Weight | 233.19 |
| Percent Purity | 99.7% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.