missing translation for 'onlineSavingsMsg'
Learn More

Bariumhydroxidemonohydrate, TRC

Product Code. 30329200
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30329200 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30329200

Brand: Toronto Research Chemicals TRCB12913810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bariumhydroxidemonohydrate
CAS 22326-55-2
Molecular Formula H2O2Ba.H2O
Quantity 10 g
InChI Formula InChI=1S/Ba.3H2O/h;3*1H2/q+2;;;/p-2
SMILES O.[OH-].[OH-].[Ba+2]
IUPAC Name barium(2+);dihydroxide;hydrate
Molecular Weight (g/mol) 189.35
Formula Weight 189.9213
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.