Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Product Code. 10156122 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Cartridge Kit
Empty Cartridges
External Booster Pump
Replacement Ultraviolet Lamp
Start-up Kit
Unit Size:
1 set
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10156122 Start-up Kit 1 set
10378343 External Booster Pump Each
10698333 Cartridge Kit Each
10433893 Empty Cartridges Each
10235983 Replacement Ultraviolet Lamp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10156122 Supplier Thermo Scientific™ Supplier No. D502132
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
In Stock
Add to basket
Add to basket
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
TRUSTED_SUSTAINABILITY
Specifications
Description Start-up Kit
For Use With (Equipment) Barnstead Easypure RoDi
Type Start-up Kit