Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Product Code. 10698333 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Cartridge Kit
Empty Cartridges
External Booster Pump
Replacement Ultraviolet Lamp
Start-up Kit
Unit Size:
1 set
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10698333 Cartridge Kit Each
10378343 External Booster Pump Each
10156122 Start-up Kit 1 set
10433893 Empty Cartridges Each
10235983 Replacement Ultraviolet Lamp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10698333 Supplier Thermo Scientific™ Supplier No. D502133
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
TRUSTED_SUSTAINABILITY
Specifications
Description Cartridge Kit
For Use With (Equipment) Barnstead Easypure RoDi
Type Cartidge Kit