Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Product Code. 10034781 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Calibration Module
Deionized Water Feed (Low Organics, Type I Water)
Deionized Water Feed (Ultralow Organics, Type I Water)
Low Water Level Pump Protector
R/O and Distilled Feed (Low Organics, Type I Water)
R/O and Distilled Feed (Ultralow Organics, Type I Water)
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10034781 R/O and Distilled Feed (Ultralow Organics, Type I Water) Each
10024771 Deionized Water Feed (Ultralow Organics, Type I Water) Each
10186251 R/O and Distilled Feed (Low Organics, Type I Water) Each
10739692 Deionized Water Feed (Low Organics, Type I Water) Each
12740896 Low Water Level Pump Protector Each
10688333 Calibration Module Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10034781 Supplier Thermo Scientific™ Supplier No. D50280
In Stock
Add to basket
Add to basket
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Nanopure Water Purifications System
Type R/O and Distilled Feed (Ultralow Organics, Type I Water)