Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
209.00 € - 1050.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10034781
|
Thermo Scientific™
D50280 |
R/O and distilled feed; Ultralow organics, type I water |
1050.00 €
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
10024771
|
Thermo Scientific™
D50281 |
Deionized water feed ; Ultralow organics, type I water |
1050.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10186251
|
Thermo Scientific™
D50282 |
R/O and distilled feed; Low organics, type I water |
1050.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10739692
|
Thermo Scientific™
D50283 |
Deionized water feed; Low organics, type I water |
1050.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10688333
|
Thermo Scientific™
E896X5 |
Calibration Module |
209.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
12740896
|
Thermo Scientific™
D8964 |
Low water level pump protector |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
- Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter